Learn More
Bio Rad Laboratories Sealing frame, helps hold heat sealing films and foils in position, for use with PX1 PCR plate sealer
Catalog No. NC2946633
Encompass
The sealing frame helps hold heat sealing films and foils in position when using the PX1 PCR plate sealer. This sealing aid prevents heat seals from shifting during the thermal sealing process and ensures complete and uniform sealing.Key FeaturesEngineered for optimal fit and function with the PX1 PCR plate sealerCompatible with many different heat seals and microplatesSturdy stainless steel design
Catalog No. NC2946633 Supplier Bio Rad Laboratories Supplier No. 1814080
The sealing frame helps hold heat sealing films and foils in position when using the PX1 PCR plate sealer. This sealing aid prevents heat seals from shifting during the thermal sealing process and ensures complete and uniform sealing.Key FeaturesEngineered for optimal fit and function with the PX1 PCR plate sealerCompatible with many different heat seals and microplatesSturdy stainless steel design