Learn More
Perkin Elmer US LLC Sealing Insert for Volatile Sample Pans
Catalog No. 502104151
Encompass_Preferred
The Universal Crimper Press requires Insert (B0144637) for Volatile Sample Pans (02190062, 02190080, and N5190788), Insert (B0505340) for Stainless Steel Pans (03190218 and 03190029), Insert (B0508921) for Standard Aluminum Pans (02190041), and Insert (B0139033) for hermetically-sealed 10, 30, and 50 µL sample pans.
Catalog No. 50-210-4151 Supplier Perkin Elmer US LLC Supplier No. B0144637
The Universal Crimper Press requires Insert (B0144637) for Volatile Sample Pans (02190062, 02190080, and N5190788), Insert (B0505340) for Stainless Steel Pans (03190218 and 03190029), Insert (B0508921) for Standard Aluminum Pans (02190041), and Insert (B0139033) for hermetically-sealed 10, 30, and 50 µL sample pans.