Learn More
Electron Microscopy Sciences Sealing Labels PetriSEAL, Red, 0.50" 108 FT
SDP
Catalog No. 501899343
Encompass_Preferred
Ideal sealing tapes for large containers and bottles to cover wider gaps ensuring a tight seal. They stretch, conform, and seal the irregular perimeter of microplates or Petri dishes. Resist most solvents and caustic agents. Seal remains flexible in moderate freezer temperatures and withstands intermittent exposure to 100 degree C.
Color: Red
Width: 0.50"
Catalog No. 50-189-9343 Supplier Electron Microscopy Sciences Supplier No. 70500R
Ideal sealing tapes for large containers and bottles to cover wider gaps ensuring a tight seal. They stretch, conform, and seal the irregular perimeter of microplates or Petri dishes. Resist most solvents and caustic agents. Seal remains flexible in moderate freezer temperatures and withstands intermittent exposure to 100 degree C.
Color: Red
Width: 0.50"