Learn More
Alkali Scientific ASI™ 96 Well Round Silicone Sealing Mat, Autoclavable, 2.0ml, 10/pk, 50/cs
SDP
Catalog No. 502335792
Encompass_Preferred
ASI™ 96 Well Sterile Silicone Sealing Mat, 50/case are designed for 96-well storage plates and are available in autoclavable format. The square capped mats are made of thermoplastic elastomer and are suitable for long-term storage. They are molded from a proprietary thermoplastic elastomer for a flexible design, have alphanumeric grid-referencing, and are autoclavable. They are solvent resistant (including DMSO, Ethanol, and Isopropanol), and suitable for UV irradiation and Gamma iIrradiation. 6N required for piercing. The temperature range is −20°C to +55°C.
Catalog No. 50-233-5792 Supplier Alkali Scientific Supplier No. MPD52S
ASI™ 96 Well Sterile Silicone Sealing Mat, 50/case are designed for 96-well storage plates and are available in autoclavable format. The square capped mats are made of thermoplastic elastomer and are suitable for long-term storage. They are molded from a proprietary thermoplastic elastomer for a flexible design, have alphanumeric grid-referencing, and are autoclavable. They are solvent resistant (including DMSO, Ethanol, and Isopropanol), and suitable for UV irradiation and Gamma iIrradiation. 6N required for piercing. The temperature range is −20°C to +55°C.