Learn More
Chemglass Life Sciences Sealing Mat, Silicone/PTFE, 6mm Plugs
SDP
Catalog No. 501539698
Encompass_Preferred
Standard SBS (7mm round diameter plug) cap mat; made with 100% premium silicone with PTFE spray coating, available in packs of 5, non-slit.
Catalog No. 50-153-9698 Supplier Chemglass Life Sciences Supplier No. CG191404
Standard SBS (7mm round diameter plug) cap mat; made with 100% premium silicone with PTFE spray coating, available in packs of 5, non-slit.