Learn More
Microsolv Technology Corporation Sealing Mat, Pre-Slit Silicone Rubber, for 96 Well Plates, Square Wells. 10/PK.
Catalog No. 502270502
Encompass_Preferred
This Sealing Mat is made of high purity Silicone Rubber for 96 Well Plates. Each well is Pre-Slit for easier penetration by Auto-injectors. The Wells are Square.
This item is not returnable. View return policy
Catalog No. 50-227-0502 Supplier Microsolv Technology Corporation Supplier No. 9509658K
This Sealing Mat is made of high purity Silicone Rubber for 96 Well Plates. Each well is Pre-Slit for easier penetration by Auto-injectors. The Wells are Square.