Learn More
Chemglass Life Sciences Sealing Mat, Silicone/PTFE, 6mm Plugs, Pre-Slit
SDP
Catalog No. 501539699
Encompass_Preferred
Standard SBS (7mm round diameter plug) cap mat; made with 100% premium silicone with PTFE spray coating, available in packs of 5, pre-slit.
Catalog No. 50-153-9699 Supplier Chemglass Life Sciences Supplier No. CG191405
Standard SBS (7mm round diameter plug) cap mat; made with 100% premium silicone with PTFE spray coating, available in packs of 5, pre-slit.