Learn More
Phenomenex Inc Silicone
Catalog No. 502400457 Shop All Phenomenex Products
Encompass_Preferred
Sealing Mats, Pierceable, 96-Round Well 8mm AH0-8633
Catalog No. 50-240-0457 Supplier Phenomenex Inc Supplier No. AH08633
Sealing Mats, Pierceable, 96-Round Well 8mm AH0-8633