Learn More
Hampton Research Gold Film Sealing Paddle
SDP
Catalog No. NC1588068
Encompass
An alternative to the plate roller or brayer for pressing films to assure a secure uniform seal around all wells on 96-well, 384-well, and 1536-well plates. These paddles are especially recommended for sealing films on raised-rim plates because they fit within the rim of the plate. Gold, film sealing paddle, plastic, nylon reinforced edge, more firm than white paddle. 1 paddle per pack.
Catalog No. NC1588068 Supplier Hampton Research Supplier No. HR4525
An alternative to the plate roller or brayer for pressing films to assure a secure uniform seal around all wells on 96-well, 384-well, and 1536-well plates. These paddles are especially recommended for sealing films on raised-rim plates because they fit within the rim of the plate. Gold, film sealing paddle, plastic, nylon reinforced edge, more firm than white paddle. 1 paddle per pack.