Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO SEALING RING 12MM FOR ADAPTER

Catalog No. 502741473 Shop All METTLER TOLEDO Products
Encompass

Sealing ring 12mm for adapter PTFE

Catalog No. 50-274-1473 Supplier METTLER TOLEDO Supplier No. 51162765
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sealing ring 12mm for adapter PTFE

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.