Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO SEALING RING 8MM PTFE FOR 3006

Catalog No. 502741797 Shop All METTLER TOLEDO Products
Encompass

Sealing ring 8mm for adapter PTFE

Catalog No. 50-274-1797 Supplier METTLER TOLEDO Supplier No. 30064134
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sealing ring 8mm for adapter PTFE

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.