Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO SEALING RING INNER 6MM PTF

Catalog No. 502741482 Shop All METTLER TOLEDO Products
Encompass

Sealing ring 6mm for adapter PTFE

Catalog No. 50-274-1482 Supplier METTLER TOLEDO Supplier No. 51162716
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sealing ring 6mm for adapter PTFE

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.