Learn More
Bio Rad Laboratories SEALING ROLLER
Catalog No. 509010046
Encompass
Roller, Sealing; Bio-Rad; For film seals; Firmly seats PCR plate sealers; Also useful with other types of PCR plate sealers, including Microseal B clear adhesive seals and Microseal F foil
Catalog No. 50-901-0046 Supplier Bio Rad Laboratories Supplier No. MSR0001
Roller, Sealing; Bio-Rad; For film seals; Firmly seats PCR plate sealers; Also useful with other types of PCR plate sealers, including Microseal B clear adhesive seals and Microseal F foil