Learn More
Chemglass Life Sciences Glassblowers Fritted Tube, Straight, Frit 50mm Diameter, Coarse Porosity, Tube O.D./54mm
SDP
Catalog No. CG20219
Encompass_Preferred
Glassblowers Fritted Tube, Straight, Frit 50mm Diameter, Coarse Porosity, Tube O.D./54mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.
Catalog No. CG20219 Supplier Chemglass Life Sciences Supplier No. CG20219
Glassblowers Fritted Tube, Straight, Frit 50mm Diameter, Coarse Porosity, Tube O.D./54mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.