Learn More
Chemglass Life Sciences Glassblowers Fritted Tube, Straight, Frit 10mm Diameter, Coarse Porosity, Tube O.D./13mm
SDP
Catalog No. CG20203
Encompass_Preferred
Glassblowers Fritted Tube, Straight, Frit 10mm Diameter, Coarse Porosity, Tube O.D./13mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.
Catalog No. CG20203 Supplier Chemglass Life Sciences Supplier No. CG20203
Glassblowers Fritted Tube, Straight, Frit 10mm Diameter, Coarse Porosity, Tube O.D./13mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.