Learn More
Andwin Scientific AluminaSeal Microplate Sealing, 100/pk
SDP
Catalog No. NC1660884
Encompass_Preferred
Ideal for cold storage, moderate and high temperature incubation. Protection of light sensitive samples and high throughput samples. Strong 2.7 mil thick pierceable aluminum film. Non-cytotoxic and sterile, temperature stable from -80 degree C to 110 degree C.
Catalog No. NC1660884 Supplier Andwin Scientific Supplier No. ALUM1000
Ideal for cold storage, moderate and high temperature incubation. Protection of light sensitive samples and high throughput samples. Strong 2.7 mil thick pierceable aluminum film. Non-cytotoxic and sterile, temperature stable from -80 degree C to 110 degree C.