Learn More
Andwin Scientific Titer Tops 100/pk
SDP
Catalog No. NC1659359
Encompass_Preferred
Multipurpose pre-cut adhesive polyethylene film. Durable and thick (4.4mil) yet easily pierced with a pipet tip, Polyethylene film that provides excellent adhesion to plastic microplates, Non-cytotoxic, sterile, solvent resistant and waterproof, Chemically resistant and thermally stable, UV transparent down to 25nm.
This item is not returnable. View return policy
Catalog No. NC1659359 Supplier Andwin Scientific Supplier No. TTOPS100
Multipurpose pre-cut adhesive polyethylene film. Durable and thick (4.4mil) yet easily pierced with a pipet tip, Polyethylene film that provides excellent adhesion to plastic microplates, Non-cytotoxic, sterile, solvent resistant and waterproof, Chemically resistant and thermally stable, UV transparent down to 25nm.