Learn More
Research Products International Corp SealMate Adhesive Sealing Dispenser Kit with 2 SealPlate Film Rolls, Sterile
SDP
Catalog No. 501977492
Encompass
SealMate Adhesive Sealing Film system features a unique method of applying sealing film on plates. SealMate provides fast, efficient and reliable seal on a plate without contamination. Film is available in roll format and reduces waste since it is supplied without protective backing or liner. Dispenser also functions as sealing paddle.
Kit is supplied with 2 rolls of film and dispenser. SealMate film minimizes evaporation, prevents contamination between well, and products secure seal. Film is stable at -40° to +120°C. Roll contains film to cover 100 plates.
SealPlate and SealMate are Trademarks of Excel Scientific, Inc.
Ideal for ELISA/EIA, Incubation, General Use
Convenient film dispensing
3X Faster sealing than individual sheets
Reduce waste - no protective liner to remove
Catalog No. 50-197-7492 Supplier Research Products International Corp Supplier No. 202552
SealMate Adhesive Sealing Film system features a unique method of applying sealing film on plates. SealMate provides fast, efficient and reliable seal on a plate without contamination. Film is available in roll format and reduces waste since it is supplied without protective backing or liner. Dispenser also functions as sealing paddle.
Kit is supplied with 2 rolls of film and dispenser. SealMate film minimizes evaporation, prevents contamination between well, and products secure seal. Film is stable at -40° to +120°C. Roll contains film to cover 100 plates.
SealPlate and SealMate are Trademarks of Excel Scientific, Inc.
Ideal for ELISA/EIA, Incubation, General Use
Convenient film dispensing
3X Faster sealing than individual sheets
Reduce waste - no protective liner to remove