Learn More
Research Products International Corp SealMate Adhesive Sealing Dispenser Kit with 2 AeraSeal Film Rolls, Sterile
SDP
Catalog No. 50998796
Encompass
SealMate Adhesive Sealing Film system features a unique method of applying sealing film on plates. SealMate provides fast, efficient and reliable seal on a plate without contamination. Film is available in roll format and reduces waste since it is supplied without protective backing or liner. Dispenser also functions as sealing paddle.
Each kit is supplied with 2 rolls of film. AeraSeal breathable sealing film is ideal for cell culture applications. Pierceable film fits culture plates or 96 well assay plates. Film is stable at -20° to +80°C. Roll contains film to cover 50 plates.
AeraSeal SealMate is a Trademark for Excel Scientific, Inc.
Catalog No. 50-998-796 Supplier Research Products International Corp Supplier No. 202550
SealMate Adhesive Sealing Film system features a unique method of applying sealing film on plates. SealMate provides fast, efficient and reliable seal on a plate without contamination. Film is available in roll format and reduces waste since it is supplied without protective backing or liner. Dispenser also functions as sealing paddle.
Each kit is supplied with 2 rolls of film. AeraSeal breathable sealing film is ideal for cell culture applications. Pierceable film fits culture plates or 96 well assay plates. Film is stable at -20° to +80°C. Roll contains film to cover 50 plates.
AeraSeal SealMate is a Trademark for Excel Scientific, Inc.