Learn More
Sigma Aldrich Fine Chemicals Biosciences SealPlate film, 100/pk
Catalog No. NC0665794
Encompass
Sheets pre-cut to fit standard multiwell plates. Both film and adhesive are inert and compatible with microplate procedures. Adhesive forms a tight, waterproof seal, preventing cross-contamination and evaporation. SealPlate film is 2 mil (0.05 mm) polyester, with a functional temperature range of -40 to +120 °C.
Catalog No. NC0665794 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z369659100EA
Sheets pre-cut to fit standard multiwell plates. Both film and adhesive are inert and compatible with microplate procedures. Adhesive forms a tight, waterproof seal, preventing cross-contamination and evaporation. SealPlate film is 2 mil (0.05 mm) polyester, with a functional temperature range of -40 to +120 °C.