Learn More
Research Products International Corp Plate Sealing Films, SealPlate, Clear, Sterile, 100 per Package
SDP
Catalog No. 50550230
Encompass
Adhesive films are specially designed for use as microplate covers. SealPlate 50 µm thickness polyester sealing films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal, not just a cover. Secure sealing of all wells eliminates "edge effects" in sensitive ELISA assays. SealPlate sealing films can also be used with tissue-culture plates for short-term storage, incubation, and containment of biohazards. SealPlate sealing films are non-pierceable. Each SealPlate sealing film measures 79.4 x 146.1 mm. SealPlate is a Trademark of Excel Scientific, Inc.
Catalog No. 50-550-230 Supplier Research Products International Corp Supplier No. 202503
Adhesive films are specially designed for use as microplate covers. SealPlate 50 µm thickness polyester sealing films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal, not just a cover. Secure sealing of all wells eliminates "edge effects" in sensitive ELISA assays. SealPlate sealing films can also be used with tissue-culture plates for short-term storage, incubation, and containment of biohazards. SealPlate sealing films are non-pierceable. Each SealPlate sealing film measures 79.4 x 146.1 mm. SealPlate is a Trademark of Excel Scientific, Inc.