Learn More
Research Products International Corp SealPlate MiniStrips, Sterile, 3 1/2 x 3/4 Inches x 2 ml Thick, 200 per Package
SDP
Catalog No. 50998377
Encompass
Adhesive film is ideal for selectively sealing and protecting one or two 8-well plate rows. Convenient MiniStrips can be used as a secondary seal to cover films that have been previously pierced. Thermo stable film forms a superior seal around each well. Perforated tabs may be removed or sealed around plate edge. Polyester based strips withstand -40°C to +120°C and are resistant to chemicals and solvents. SealPlate MiniStrips is a Trademark of Excel Scientific, Inc.
Catalog No. 50-998-377 Supplier Research Products International Corp Supplier No. 202507
Adhesive film is ideal for selectively sealing and protecting one or two 8-well plate rows. Convenient MiniStrips can be used as a secondary seal to cover films that have been previously pierced. Thermo stable film forms a superior seal around each well. Perforated tabs may be removed or sealed around plate edge. Polyester based strips withstand -40°C to +120°C and are resistant to chemicals and solvents. SealPlate MiniStrips is a Trademark of Excel Scientific, Inc.