Learn More
Research Products International Corp SealPlate Rolls for SealMate DIS Kit, Non-S, 2/pk
SDP
Catalog No. 502230224
Encompass
Replacement rolls for use with the SealMate Dispenser Kit.
SealPlate films provide a fast, efficient and reliable seal on a plate without contamination. Film is available in roll format and reduces waste since it is supplied without protective backing or liner. SealMate film minimizes evaporation and prevents contamination between wells. Film is stable at -40°C to +120°C. SealPlate rolls contains enough film to cover 100 plates.
SealPlate and SealMate are Trademarks of Excel Scientific, Inc.
Convenient film dispensing
3X Faster sealing than individual sheets
Reduce waste - no protective liner to remove
Catalog No. 50-223-0224 Supplier Research Products International Corp Supplier No. 202553NS
Replacement rolls for use with the SealMate Dispenser Kit.
SealPlate films provide a fast, efficient and reliable seal on a plate without contamination. Film is available in roll format and reduces waste since it is supplied without protective backing or liner. SealMate film minimizes evaporation and prevents contamination between wells. Film is stable at -40°C to +120°C. SealPlate rolls contains enough film to cover 100 plates.
SealPlate and SealMate are Trademarks of Excel Scientific, Inc.
Convenient film dispensing
3X Faster sealing than individual sheets
Reduce waste - no protective liner to remove