Learn More
Bellco Glass, Inc. SealPlateSterile Film,2mil For Microplates
SDP
Catalog No. 501945600
Encompass_Preferred
Sterile sealing films prevent evaporation and protect samples during processing, storage, and shipment. They have superior quality film and uniform adhesive. With 2 perforated end tabs for accurate positioning. Removable end-tab backing for sealing around ends of plate. Films are packaged in zipper bags. Manufactured from 2mil polyester. 80x146, -40C to 120C, ELISA, Co2 exchange-no, Pierceable-No, DN/RNase free-No
Catalog No. 50-194-5600 Supplier Bellco Glass, Inc. Supplier No. 440000200
Sterile sealing films prevent evaporation and protect samples during processing, storage, and shipment. They have superior quality film and uniform adhesive. With 2 perforated end tabs for accurate positioning. Removable end-tab backing for sealing around ends of plate. Films are packaged in zipper bags. Manufactured from 2mil polyester. 80x146, -40C to 120C, ELISA, Co2 exchange-no, Pierceable-No, DN/RNase free-No