Learn More
Sigma Aldrich Fine Chemicals Biosciences SealPlateR film sterile
Catalog No. 502900352
Encompass
Sheets pre-cut to fit standard multiwell plates. Both film and adhesive are inert and compatible with microplate procedures. Adhesive forms a tight waterproof seal preventing cross-contamination and evaporation. SealPlate film is 2Amil (0.05Amm) polyester with a functional temperature range of -40Ato 120AC.
Catalog No. 50-290-0352 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z369667100EA
Sheets pre-cut to fit standard multiwell plates. Both film and adhesive are inert and compatible with microplate procedures. Adhesive forms a tight waterproof seal preventing cross-contamination and evaporation. SealPlate film is 2Amil (0.05Amm) polyester with a functional temperature range of -40Ato 120AC.