Learn More
USA Scientific Inc Seal-Rite .5mL centri tube
Catalog No. 502745208
Encompass
Seal-Rite 0.5 mL graduated microcentrifuge tube, natural, polypropylene, 5 bags of 200 (1000 tubes)
This item is not returnable. View return policy
Catalog No. 50-274-5208 Supplier USA Scientific Inc Supplier No. 16050000
Seal-Rite 0.5 mL graduated microcentrifuge tube, natural, polypropylene, 5 bags of 200 (1000 tubes)