Learn More
MSC SEALS
Catalog No. 5000687341
Encompass
These seals are designed for strapping applications, providing secure and reliable closure for 0.75 in. wide straps.
  • Manufactured from the finest raw materials
  • Known for quality, consistency, and reliability
  • Complies with ASTM Standard D3950 for open and closed seals
  • Made of steel
  • Thread on, open seal type
Catalog No. 50-006-87341 Supplier MSC Supplier No. 01700863
These seals are designed for strapping applications, providing secure and reliable closure for 0.75 in. wide straps.
  • Manufactured from the finest raw materials
  • Known for quality, consistency, and reliability
  • Complies with ASTM Standard D3950 for open and closed seals
  • Made of steel
  • Thread on, open seal type