Learn More
MSC SEALS
Catalog No. 5000677406
Encompass
These seals are designed for use with steel strapping, offering high joint efficiency. They are manufactured from precision-tested galvanized steel and comply with federal government standards and ASTM Standard D3953.
  • Made from precision-tested galvanized steel
  • Engineered for high joint efficiency with regular duty and heavy-duty steel
  • Meet federal government standards and ASTM Standard D3953
Catalog No. 50-006-77406 Supplier MSC Supplier No. 01700798
These seals are designed for use with steel strapping, offering high joint efficiency. They are manufactured from precision-tested galvanized steel and comply with federal government standards and ASTM Standard D3953.
  • Made from precision-tested galvanized steel
  • Engineered for high joint efficiency with regular duty and heavy-duty steel
  • Meet federal government standards and ASTM Standard D3953