Learn More
Ace Glass, Inc. Seals, aluminum, 20mm, pressure release, PTFE/silicone liner, pk/100.
SDP
Catalog No. 50978413
Encompass
Seals, aluminum, 20mm, pressure release, PTFE/silicone liner, pk/100.
Catalog No. 50-978-413 Supplier Ace Glass, Inc. Supplier No. 570833
Seals, aluminum, 20mm, pressure release, PTFE/silicone liner, pk/100.