Learn More
Ace Glass, Inc. Seals, aluminum, 20mm, pressure release, PTFE/molded butyl rubber liner, cs/1000
SDP
Catalog No. 5097215
Encompass
Seals, aluminum, 20mm, pressure release, PTFE/molded butyl rubber liner, cs/1000
Catalog No. 50-972-15 Supplier Ace Glass, Inc. Supplier No. 570812
Seals, aluminum, 20mm, pressure release, PTFE/molded butyl rubber liner, cs/1000