Learn More
Ace Glass, Inc. Seals, aluminum, 20mm, PTFE/molded butyl rubber liner, cs/1000
SDP
Catalog No. 5097213
Encompass
Seals, aluminum, 20mm, PTFE/molded butyl rubber liner, cs/1000
Catalog No. 50-972-13 Supplier Ace Glass, Inc. Supplier No. 570806
Seals, aluminum, 20mm, PTFE/molded butyl rubber liner, cs/1000