Learn More
Ace Glass, Inc. Seals, aluminum, 20mm crimp, PTFE/silicone liner, cs/1000
SDP
Catalog No. 50971997
Encompass
20mm aluminum crimp seals for Type 1, heavy wall, clear borosilicate microwave digestion vials (vial part # 50971996, 50978405, NC0827007). Open top with 0.125 thick PTFE-faced silicone septa. Case/1000.
Catalog No. 50-971-997 Supplier Ace Glass, Inc. Supplier No. 570510
20mm aluminum crimp seals for Type 1, heavy wall, clear borosilicate microwave digestion vials (vial part # 50971996, 50978405, NC0827007). Open top with 0.125 thick PTFE-faced silicone septa. Case/1000.