Learn More
Alkali Scientific SealSafe MiniStrips™ Sealing Film, Sterile, 200/pk
SDP
Catalog No. 502388937
Encompass_Preferred
SealSafe MiniStrips™ sealing films are designed for sealing only one or two 8-well rows at a time on either strip-well or standard plates whenever rows must be selectively protected or accessed. Each MiniStrips film is 19.0 x 102.9mm, including 2 positioning tabs. The long dimension with the tabs removed is 82.6mm. MiniStrips are supplied in sheets of four from which strips can be peeled individually or two-at a time for application to plates. Sterile product is packaged in 100 um tamper-evident bags of 50 sheets of 4 sealing film strips.
Catalog No. 50-238-8937 Supplier Alkali Scientific Supplier No. SF22MLS
SealSafe MiniStrips™ sealing films are designed for sealing only one or two 8-well rows at a time on either strip-well or standard plates whenever rows must be selectively protected or accessed. Each MiniStrips film is 19.0 x 102.9mm, including 2 positioning tabs. The long dimension with the tabs removed is 82.6mm. MiniStrips are supplied in sheets of four from which strips can be peeled individually or two-at a time for application to plates. Sterile product is packaged in 100 um tamper-evident bags of 50 sheets of 4 sealing film strips.