Learn More
Alkali Scientific SealSafe RollRight™, Dispenser +1 Roll of SealSafe polyester Film, 500 Pre-Scored Sheets
SDP
Catalog No. 502389327
Encompass_Preferred
Self-standing, non-slip supports hold the pre-scored polyester film roll securely in place for quick and easy bench-top dispensement. The roll unwind tension can be easily adjusted to user preference. Continuous backing liner can be torn off or fed directly into a waste basket for disposal. SealSafe is economial and facilitates quicker, more efficient sealing. It provides a secure seal and is easy to remove. Rigid polyester film features good clarity and easy handling. Can be used as a temporary seal while awaiting further processing.
Catalog No. 50-238-9327 Supplier Alkali Scientific Supplier No. SF23ML
Self-standing, non-slip supports hold the pre-scored polyester film roll securely in place for quick and easy bench-top dispensement. The roll unwind tension can be easily adjusted to user preference. Continuous backing liner can be torn off or fed directly into a waste basket for disposal. SealSafe is economial and facilitates quicker, more efficient sealing. It provides a secure seal and is easy to remove. Rigid polyester film features good clarity and easy handling. Can be used as a temporary seal while awaiting further processing.