Learn More
Alkali Scientific SealSafe Titan™ Sealing Film Rolls, 2 Rolls/pk, 100 Sheets/Roll, Sterile
SDP
Catalog No. 502388977
Encompass_Preferred
SealSafe's dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Cost-effective replacement rolls are sold separately. Ideal for use under the hood; the SealMate dispenser minimizes film contamination. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 72% less material than SealPlate sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded green band. (This item is for 2 replacement rolls only. For stater kit, order SKU SF25ML or SF25ML-S for sterile version.)
Catalog No. 50-238-8977 Supplier Alkali Scientific Supplier No. SF24MLS
SealSafe's dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Cost-effective replacement rolls are sold separately. Ideal for use under the hood; the SealMate dispenser minimizes film contamination. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 72% less material than SealPlate sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded green band. (This item is for 2 replacement rolls only. For stater kit, order SKU SF25ML or SF25ML-S for sterile version.)