Learn More
Alkali Scientific SealSafe Titan™ Sealing Film Starter Kit, Titan Dispenser + 2 Rolls of SealSafe Film, 100 Sheets/Roll
SDP
Catalog No. 502389229
Encompass_Preferred
SealSafe's dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Cost-effective replacement rolls are sold separately. Ideal for use under the hood; the SealMate dispenser minimizes film contamination. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 72% less material than SealPlate sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded green band.
Catalog No. 50-238-9229 Supplier Alkali Scientific Supplier No. SF25ML
SealSafe's dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Cost-effective replacement rolls are sold separately. Ideal for use under the hood; the SealMate dispenser minimizes film contamination. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 72% less material than SealPlate sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded green band.