Learn More
Grainger Bessey Specialty Clamp, Universal, With Suction Cup, 1 ea
Catalog No. 502785287
Encompass_Preferred
This universal specialty clamping tool is designed for precision solid surface installations. It features an integrated suction cup to securely hold materials during fabrication and assembly tasks.
  • Features integrated suction cup for secure material positioning
  • Designed for specialized solid surface installation applications
  • Provides 48.0 lbs of clamping force
  • Equipped with a 4.0 in. handle length
  • Constructed for durability with a lifetime warranty against manufacturer defects
Catalog No. 50-278-5287 Supplier Grainger Supplier No. 19C756
This universal specialty clamping tool is designed for precision solid surface installations. It features an integrated suction cup to securely hold materials during fabrication and assembly tasks.
  • Features integrated suction cup for secure material positioning
  • Designed for specialized solid surface installation applications
  • Provides 48.0 lbs of clamping force
  • Equipped with a 4.0 in. handle length
  • Constructed for durability with a lifetime warranty against manufacturer defects