Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Seamless Cellulose Dialysis Tubing
SCI_ED

Catalog No. S413002 Shop All Aldon Products
Change view
Click to view available options
Diameter (Metric):
15.9 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S413002 15.9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S413002 Supplier Innovating Science™ Supplier No. DD010610FT

missing translation for 'validMainframeMsg'

Retains materials with 12,000 and higher molecular weight.

  • Especially useful in analytical chemistry, biology, and pharmaceutical fields
  • Highly permeable to water and low molecular weight compounds
  • Contains glycerin, water, 1% sulfur
  • Must be stored in air-tight container or rehumidified before use
  • Pore diameter: 4.8nm

† Average values.

Specifications

Diameter (Metric) 15.9 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.