Learn More
VAZYME INTERNATIONAL LLC Vazyme Cloning Kit, Ultra One Step, 1–11 Fragments, High Efficiency
Catalog No. NC3979690
Encompass
The ClonExpress Ultra One Step Cloning Kit V3 uses optimized technology for highly efficient homologous recombination of 1 to 11 DNA fragments. It features a newly optimized 2 × CE Mix V3, which ensures a high cloning success rate even with complicated situations like large vectors and GC-rich/poor homologous arms. It also enhances compatibility with varying amounts of DNA input, addressing issues of reduced cloning efficiency due to low DNA fragment concentration.
  • Robust performance: assembles 1 to 11 fragments with a positive rate exceeding 95%.
  • High efficiency: rapid recombination in 5 to 30 min, leading to high clone numbers and positivity rates.
  • Special optimization: compatible with 15 to 35 bp overlaps, not just 15 to 20 bp.
  • Broad compatibility: suitable for ultra-low DNA input, GC-rich/poor overlaps, and low-purity samples.
Catalog No. NC3979690 Supplier VAZYME INTERNATIONAL LLC Supplier No. C11701
The ClonExpress Ultra One Step Cloning Kit V3 uses optimized technology for highly efficient homologous recombination of 1 to 11 DNA fragments. It features a newly optimized 2 × CE Mix V3, which ensures a high cloning success rate even with complicated situations like large vectors and GC-rich/poor homologous arms. It also enhances compatibility with varying amounts of DNA input, addressing issues of reduced cloning efficiency due to low DNA fragment concentration.
  • Robust performance: assembles 1 to 11 fragments with a positive rate exceeding 95%.
  • High efficiency: rapid recombination in 5 to 30 min, leading to high clone numbers and positivity rates.
  • Special optimization: compatible with 15 to 35 bp overlaps, not just 15 to 20 bp.
  • Broad compatibility: suitable for ultra-low DNA input, GC-rich/poor overlaps, and low-purity samples.