Learn More
Grainger SCI Seamless Nipple, 304/304L Stainless Steel, S80, 1-1/2 in. x 3 in., 1 ea
Catalog No. 502785088
Encompass_Preferred
This seamless nipple is designed for high-pressure industrial applications, constructed from durable 304/304L stainless steel to provide excellent corrosion resistance. It features an S80 schedule and NPT connection type, ensuring a reliable, leak-resistant seal in piping systems.
  • Constructed from 304/304L stainless steel
  • Designed for high-pressure service with S80 schedule
  • Features standard NPT pipe fitting connection
  • Compliant with ASTM A312, ASTM A733, and ASME B36.19 standards
  • Meets ASME B1.20.1 specifications
  • Certified under ISO 9001:2008
Catalog No. 50-278-5088 Supplier Grainger Supplier No. 196U36
This seamless nipple is designed for high-pressure industrial applications, constructed from durable 304/304L stainless steel to provide excellent corrosion resistance. It features an S80 schedule and NPT connection type, ensuring a reliable, leak-resistant seal in piping systems.
  • Constructed from 304/304L stainless steel
  • Designed for high-pressure service with S80 schedule
  • Features standard NPT pipe fitting connection
  • Compliant with ASTM A312, ASTM A733, and ASME B36.19 standards
  • Meets ASME B1.20.1 specifications
  • Certified under ISO 9001:2008