Learn More
Grainger Anvil Smith Cooper Pipe Nipple, 304 Stainless Steel, Schedule 80, 2 in. x 6 in., 1 ea.
Catalog No. 502785098
Encompass_Preferred
This seamless pipe nipple is designed for high-pressure industrial applications, constructed from durable 304 stainless steel for reliable performance and very good corrosion resistance. The component features precision machining and is threaded on both ends for secure, leak-resistant connections.
  • Schedule 80 construction for high-pressure durability
  • Constructed from 304 stainless steel
  • Threaded on both ends with 2 in. NPT male connections
  • 6 in. overall length
  • Meets ASME B1.20.1 specifications
Catalog No. 50-278-5098 Supplier Grainger Supplier No. 196U53
This seamless pipe nipple is designed for high-pressure industrial applications, constructed from durable 304 stainless steel for reliable performance and very good corrosion resistance. The component features precision machining and is threaded on both ends for secure, leak-resistant connections.
  • Schedule 80 construction for high-pressure durability
  • Constructed from 304 stainless steel
  • Threaded on both ends with 2 in. NPT male connections
  • 6 in. overall length
  • Meets ASME B1.20.1 specifications