Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bolle Safety Seamless Vision Virtual Code Photochromic

Catalog No. 17033380
Change view
Click to view available options
Product Type:
Prescription Safety Glasses Program, Antibody
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-033-380 Prescription Safety Glasses Program, Antibody
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-033-380 Supplier Bolle Safety Supplier No. VOUCPV73U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Discover Seamless Prescription Program, the easiest way to get your tailor-made safety glasses. Seamless Vision™ offers an integrated solution, combining cutting edge technology and dedicated customer service for prescription safety eyewear.

  • Offers an integrated solution, combining cutting edge technology and dedicated customer service for prescription safety eyewear.
  • Try on frames virtually, ensuring both functionality and aesthetics.
  • Available with single vision or progressive lenses for indoor or outdoor usage, Seamless Vision™ combines advanced lens design with our user-friendly virtual platform.
  • Your prescription safety eyewear just one click away.
  • The received codes (weither virtual or on a physical single use card) enables the holder to access the Seamless Vision platform and order his/her tailored prescription safety eyewear.

Specifications

Antigen SIGLEC H
Applications Flow Cytometry
Classification Monoclonal
Clone eBio440c
Concentration 0.2 mg/mL
Conjugate APC
Product Type Prescription Safety Glasses Program, Antibody
Formulation PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Gene Siglech
Gene Symbols Siglech
Host Species Rat
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233274
Target Species Mouse
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.