Learn More
Lonza Walkersville SeaPlaque Agarose
Catalog No. 502120226
Encompass
SeaPlaque Agarose is the original low melting temperature agarose. This molecular biology grade agarose produces gels with greater sieving properties and higher clarity than standard melting temperature agarose. The low melting temperature of SeaPlaque Agarose makes it ideal for preparative DNA and RNA electrophoresis, while its low gelling temperature is ideal for cloning of tissue culture cells and viral plaque assays.Analytical Specifications:Gelling temperature (1.5%) 26°C - 30°CMelting temperature (1.5%) ≤65°CGel strength (1%) ≥200 g/cm²Information about bulk and custom packaging is available through Lonza’s Customer Service Team.Benefits:All Lonza Agarose is quality tested to certify performanceLow EEOLow gelling and melting temperatureApplications:Cloning of tissue culture cellsViral plaque assaysDNA/RNA preparative electrophoresisContent:SeaPlaque Agarose 500 g
Catalog No. 50-212-0226 Supplier Lonza Walkersville Supplier No. 50104
SeaPlaque Agarose is the original low melting temperature agarose. This molecular biology grade agarose produces gels with greater sieving properties and higher clarity than standard melting temperature agarose. The low melting temperature of SeaPlaque Agarose makes it ideal for preparative DNA and RNA electrophoresis, while its low gelling temperature is ideal for cloning of tissue culture cells and viral plaque assays.Analytical Specifications:Gelling temperature (1.5%) 26°C - 30°CMelting temperature (1.5%) ≤65°CGel strength (1%) ≥200 g/cm²Information about bulk and custom packaging is available through Lonza’s Customer Service Team.Benefits:All Lonza Agarose is quality tested to certify performanceLow EEOLow gelling and melting temperatureApplications:Cloning of tissue culture cellsViral plaque assaysDNA/RNA preparative electrophoresisContent:SeaPlaque Agarose 500 g