Learn More
ProSci Seasonal H1N1 Neuraminidase Peptide

Description
Specifications
Specifications
| Species | Influenza A Virus (H1N1) |
| Accession Number | ACA33620 |
| Common Name | neuraminidase |
| Concentration | 200 μg/mL |
| Content And Storage | Store Seasonal H1N1 Neuraminidase peptide at -20°C, stable for one year. |
| Format | Liquid |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide |
| For Use With (Application) | Blocking Assay |
| Quantity | 0.05 mg |
| Sequence | Proprietary |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.