Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Seasonal H1N1 Nonstructural Protein 1 Peptide
SDP

Catalog No. 16594864
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-594-864 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-594-864 Supplier ProSci Supplier No. 5357P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

Seasonal H1N1 Nonstructural Protein 1 peptide is meant to be used as a positive control in ELISA applications to detect Seasonal Influenza A antibody.

Specifications

Species Influenza A Virus (H1N1)
Accession Number ABP49398
Common Name nonstructural protein 1
Concentration 200 μg/mL
Content And Storage Store Seasonal H1N1 Nonstructural Protein 1 peptide (at -20°C, stable for one year.)
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol NS1
Quantity 0.05 mg
Sequence Proprietary
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.