Learn More
Glass Expansion SeaSpray DC Nebulizer 0.4mL/min with 0.25mm ID x 1.6mm OD sample tube
SDP
Catalog No. 5000556324
Encompass_Preferred
SeaSpray DC Nebulizer 0.4mL/min with 0.25mm ID x 1.6mm OD sample tube Sourced from Glass Expansion
Catalog No. 50-005-56324 Supplier Glass Expansion Supplier No. A221USS04Q
SeaSpray DC Nebulizer 0.4mL/min with 0.25mm ID x 1.6mm OD sample tube Sourced from Glass Expansion