Learn More
Glass Expansion SeaSpray DC Nebulizer 0.4mL/min, Includes NFT-16-50 UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 piece) and 70-803-1086 Gas Fitting with Ratchet Nebulizer connector
SDP
Catalog No. 501907390
Encompass_Preferred
SeaSpray DC Nebulizer 0.4 mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 piece) and DC Gas Fitting Type 12 with ratchet connector (1 piece). Compatible with Agilent 4500/7500.Optimum nebulizer gas flow rate is 1.0 L/min and sample flow rate is 0.4 mL/min. Recommended not to exceed optimum 0.4 mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 1.0 L/min gas flow rate. Can tolerate up to 20% TDS. We recommend the use of the Elegra Argon Humidifier to achieve best performance with high TDS.Replacement parts include P/Ns 502702959 and 501907288.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7390 Supplier Glass Expansion Supplier No. A121USS04
SeaSpray DC Nebulizer 0.4 mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 piece) and DC Gas Fitting Type 12 with ratchet connector (1 piece). Compatible with Agilent 4500/7500.Optimum nebulizer gas flow rate is 1.0 L/min and sample flow rate is 0.4 mL/min. Recommended not to exceed optimum 0.4 mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 1.0 L/min gas flow rate. Can tolerate up to 20% TDS. We recommend the use of the Elegra Argon Humidifier to achieve best performance with high TDS.Replacement parts include P/Ns 502702959 and 501907288.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001