Learn More
Glass Expansion SeaSpray DC Nebulizer 2mL/min, Includes NFT-075 UniFit Connector with 0.75mm ID x 1.3mm OD x 700mm long tubing (1 piece), and 70-803-1070 Gas Fitting with Ratchet Nebulizer connector
SDP
Catalog No. 501907449
Encompass_Preferred
SeaSpray DC Nebulizer 2.0 mL/min, includes UniFit Connector with 0.75mm ID x 1.3mm OD x 700mm long tubing (1 piece) and DC Gas Fitting Type 21 with ratchet connector (1 piece). Compatible with PerkinElmer Optima ICP-OES.Optimum nebulizer gas flow rate is 0.7 L/min and sample flow rate is 2.0 mL/min. Recommended not to exceed optimum 2.0 mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 0.7 L/min gas flow rate. Can tolerate up to 20% TDS. We recommend the use of the Elegra Argon Humidifier to achieve best performance with high TDS.Replacement parts include P/Ns NC1625341 and 70-803-1070.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7449 Supplier Glass Expansion Supplier No. A2107USS2
SeaSpray DC Nebulizer 2.0 mL/min, includes UniFit Connector with 0.75mm ID x 1.3mm OD x 700mm long tubing (1 piece) and DC Gas Fitting Type 21 with ratchet connector (1 piece). Compatible with PerkinElmer Optima ICP-OES.Optimum nebulizer gas flow rate is 0.7 L/min and sample flow rate is 2.0 mL/min. Recommended not to exceed optimum 2.0 mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 0.7 L/min gas flow rate. Can tolerate up to 20% TDS. We recommend the use of the Elegra Argon Humidifier to achieve best performance with high TDS.Replacement parts include P/Ns NC1625341 and 70-803-1070.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001