Learn More
Perkin Elmer US LLC SeaSpray DC Nebulizer, 2.0 mL/min
Catalog No. 502105554
Encompass_Preferred
The SeaSpray nebulizer is ideal for samples with high dissolved solids and offers outstanding nebulization efficiency for trace level analyses. It also includes the Connector U-Series 1.3 mm O.D. x 0.75 mm I.D. (N0777512). Argon Flow Rate: 0.7 L/min Uptake Rate: 2.0 mL/min Material: Glass
Catalog No. 50-210-5554 Supplier Perkin Elmer US LLC Supplier No. N0811305
The SeaSpray nebulizer is ideal for samples with high dissolved solids and offers outstanding nebulization efficiency for trace level analyses. It also includes the Connector U-Series 1.3 mm O.D. x 0.75 mm I.D. (N0777512). Argon Flow Rate: 0.7 L/min Uptake Rate: 2.0 mL/min Material: Glass