Learn More
Perkin Elmer US LLC SeaSpray Nebulizer, 0.4 mL/min
Catalog No. 502103895
Encompass_Preferred
All NexION 1000/2000 SeaSpray Nebulizers are Direct Connection (DC) and include a Gas Fitting with ratchet connector (N8151079).
Catalog No. 50-210-3895 Supplier Perkin Elmer US LLC Supplier No. N8151070
All NexION 1000/2000 SeaSpray Nebulizers are Direct Connection (DC) and include a Gas Fitting with ratchet connector (N8151079).